trans-2,5-Dimethylpiperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D179604-5G
SIALD179604-5GEA 80.7 EUR
SIALD179604-5G
trans-2,5-Dimethylpiperazin, Sigma-Aldrich®
trans-2,5-Dimethylpiperazin
Formel: C₆H₁₄N₂
Molecular Weight: 114,19 g/mol
MDL: MFCD00070516
CAS-Nummer: 2815-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR