Dipentaerythrit, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,2'-(Oxybis(methylene))bis(2-(hydroxymethyl)propane-1,3-diol)
M-Clarity™ Qualitätsstufe = MQ100
D203203-500G
SIALD203203-500GEA
82.4
EUR
SIALD203203-500G
Dipentaerythrit, Sigma-Aldrich®
Dipentaerythrit
Formel:
C₁₀H₂₂O₇ Molecular Weight: 254,28 g/mol |
MDL:
MFCD00004691 CAS-Nummer: 126-58-9 EINECS: 204-794-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...