N,N'-Diphenylbenzidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine

M-Clarity™ Qualitätsstufe = MQ100

D205206-10G
SIALD205206-10GEA 98.2 EUR
SIALD205206-10G
N,N'-Diphenylbenzidin, Sigma-Aldrich®
N,N'-Diphenylbenzidin
Formel: C₂₄H₂₀N₂
Molecular Weight: 336,44 g/mol
MDL: MFCD00003016
CAS-Nummer: 531-91-9
EINECS: 208-521-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR