trans,trans-1,4-Diphenyl-1,3-butadien, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
trans,trans-DPB, trans,trans-Bistyryl, trans,trans-PPB
M-Clarity™ Qualitätsstufe = MQ200
D206008-25G
D206008-5G
SIALD206008-25GEA
116
EUR
SIALD206008-25G
SIALD206008-5G
trans,trans-1,4-Diphenyl-1,3-butadien, Sigma-Aldrich®
trans,trans-1,4-Diphenyl-1,3-butadien
Formel:
C₁₆H₁₄ Molecular Weight: 206.29 g/mol |
MDL:
MFCD00004791 CAS-Nummer: 538-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...