2,2-Diphenylethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-2,2-diphenylethane, 2,2-Diphenylethanamine

M-Clarity™ Qualitätsstufe = MQ100

D206709-5G
SIALD206709-5GEA 41.2 EUR
SIALD206709-5G
2,2-Diphenylethylamin, Sigma-Aldrich®
2,2-Diphenylethylamin
Formel: C₁₄H₁₅N
Molecular Weight: 197,28 g/mol
MDL: MFCD00008143
CAS-Nummer: 3963-62-0
EINECS: 223-565-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR