4-(Diphenylmethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diphenyl-4-pyridylmethan, 4-Benzhydrylpyridin

M-Clarity™ Qualitätsstufe = MQ100

D213209-10G
SIALD213209-10GEA 118 EUR
SIALD213209-10G
4-(Diphenylmethyl)pyridin, Sigma-Aldrich®
4-(Diphenylmethyl)pyridine
Formel: C₁₈H₁₅N
Molecular Weight: 245.32 g/mol
CAS-Nummer: 3678-72-6
EINECS: 222-955-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR