Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff), Sigma-Aldrich®

Lieferant: Merck

D2425-100G
SIALD2425-100GEA 75.5 EUR
SIALD2425-100G
Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff), Sigma-Aldrich®
Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff)
Formel: C₉H₁₀Cl₂N₂O
Molecular Weight: 233,10 g/mol
MDL: MFCD00018136
CAS-Nummer: 330-54-1
EINECS: 206-354-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR