4,5-Diaminopyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyrimidine-4,5-diamine

M-Clarity™ Qualitätsstufe = MQ100

D24501-1G
SIALD24501-1GEA 161 EUR
SIALD24501-1G
4,5-Diaminopyrimidin, Sigma-Aldrich®
4,5-Diaminopyrimidine
Formel: C₄H₆N₄
Molecular Weight: 110,1200 g/mol
MDL: MFCD00006113
CAS-Nummer: 13754-19-3
EINECS: 237-339-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR