N,N'-Ethylendianilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

D27004-25G
SIALD27004-25GEA 422 EUR
SIALD27004-25G
N,N'-Ethylendianilin, Sigma-Aldrich®
N,N'-Ethylendianilin
Formel: C₆H₅NHCH₂CH₂NHC₆H₅
Siedepunkt: 228 °C (1013 hPa)
Schmelzpunkt: 68 °C
MDL: MFCD00003019
CAS-Nummer: 150-61-8
EINECS: 205-765-6
Merck Index: 13,03011

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR