3,4-Bis(benzyloxy)benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,4-Dibenzyloxybenzaldehyde
M-Clarity™ Qualitätsstufe = MQ100
D36003-10G
SIALD36003-10GEA
126
EUR
SIALD36003-10G
3,4-Bis(benzyloxy)benzaldehyd, Sigma-Aldrich®
3,4-Bis(benzyloxy)benzaldehyd
Formel:
C₂₁H₁₈O₃ Molecular Weight: 318,3658 g/mol |
MDL:
MFCD00004776 CAS-Nummer: 5447-02-9 EINECS: 226-662-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...