Dimetridazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,2-Dimethyl-5-nitroimidazole
M-Clarity™ Qualitätsstufe = MQ200
D4025-1KG
SIALD4025-1KGEA
0
EUR
SIALD4025-1KG
Dimetridazol, Sigma-Aldrich®
Dimetridazol
Formel:
C₅H₇N₃O₂ Molecular Weight: 141.13 g/mol |
MDL:
MFCD00047046 CAS-Nummer: 551-92-8 EINECS: 209-001-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...