Desoxybenzoin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Phenylacetophenone, 1,2-Diphenylethanone

M-Clarity™ Qualitätsstufe = MQ100

D4369-25G
SIALD4369-25GEA 43.2 EUR
SIALD4369-25G
Desoxybenzoin, Sigma-Aldrich®
Desoxybenzoin
Formel: C₁₄H₁₂O
Molecular Weight: 196,25 g/mol
MDL: MFCD00003081
CAS-Nummer: 451-40-1
EINECS: 207-193-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR