Dexamethason, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum
M-Clarity™ Qualitätsstufe = MQ200
D4902-100MG
D4902-1G
D4902-25MG
D4902-500MG
SIALD4902-100MGEA
102
EUR
SIALD4902-100MG
SIALD4902-1G
SIALD4902-25MG
SIALD4902-500MG
Dexamethason, Sigma-Aldrich®
Dexamethason
Formel:
C₂₂H₂₉FO₅ Schmelzpunkt: 268…271 °C |
MDL:
MFCD00064136 CAS-Nummer: 50-02-2 EINECS: 200-003-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...