1,8-Dichlor-9,10-anthrachinon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

D56403-25G D56403-100G
SIALD56403-25GEA 67.6 EUR
SIALD56403-25G SIALD56403-100G
1,8-Dichlor-9,10-anthrachinon, Sigma-Aldrich®
1,8-Dichlor-9,10-anthrachinon
Formel: C₁₄H₆Cl₂O₂
MDL: MFCD00001191
CAS-Nummer: 82-43-9
EINECS: 201-420-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR