4,5-Dichlor-2-nitroanilin ≥96%, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
D68002-10G
SIALD68002-10GEA
166
EUR
SIALD68002-10G
4,5-Dichlor-2-nitroanilin ≥96%, Sigma-Aldrich®
4,5-Dichlor-2-nitroanilin
Formel:
C₆H₄Cl₂N₂O₂ Molecular Weight: 207,02 g/mol Schmelzpunkt: 177…179 °C (Lit.) |
MDL:
MFCD00007770 CAS-Nummer: 6641-64-1 EINECS: 229-657-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...