2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Deoxyguanosine triphosphate trisodium salt, 2'-Deoxyguanosine-5'-triphosphate trisodium salt

D7170-.5ML
SIALD7170-.5MLEA 44.8 EUR
SIALD7170-.5ML
2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3), Sigma-Aldrich®
2'-Desoxyguanosin-5'-triphosphat Trinatriumsalz (dGTP-Na3)
Formel: C₁₀H₁₃N₅Na₃O₁₃P₃
Molecular Weight: 573,12 g/mol
MDL: MFCD00080337
CAS-Nummer: 93919-41-6
EINECS: 300-026-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR