2'-Desoxyadenosin Monohydrat, Sigma-Aldrich®
Lieferant: Merck
D7400-250MG
D7400-500MG
D7400-1G
D7400-25G
D7400-5G
SIALD7400-250MGEA
56.8
EUR
SIALD7400-250MG
SIALD7400-500MG
SIALD7400-1G
SIALD7400-25G
SIALD7400-5G
2'-Desoxyadenosin Monohydrat, Sigma-Aldrich®
2'-Desoxyadenosin Monohydrat
Formel:
C₁₀H₁₃N₅O₃·1H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149364 CAS-Nummer: 16373-93-6 EINECS: 213-488-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...