2'-Desoxyadenosin Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

D7400-250MG D7400-500MG D7400-1G D7400-25G D7400-5G
SIALD7400-250MGEA 54.1 EUR
SIALD7400-250MG SIALD7400-500MG SIALD7400-1G SIALD7400-25G SIALD7400-5G
2'-Desoxyadenosin Monohydrat, Sigma-Aldrich®
2'-Desoxyadenosin Monohydrat
Formel: C₁₀H₁₃N₅O₃·1H₂O
Molecular Weight: 269.26 g/mol (251.24 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149364
CAS-Nummer: 16373-93-6
EINECS: 213-488-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR