2',5'-Didesoxyadenosin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

D7408-5MG
SIALD7408-5MGEA 255 EUR
SIALD7408-5MG
2',5'-Didesoxyadenosin, Sigma-Aldrich®
2',5'-Didesoxyadenosin
Formel: C₁₀H₁₃N₅O₂
MDL: MFCD00210904
CAS-Nummer: 6698-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR