1,4-Dicyan-2-buten, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydromucononitrile

M-Clarity™ Qualitätsstufe = MQ100

D77206-5G
SIALD77206-5GEA 59.4 EUR
SIALD77206-5G
1,4-Dicyan-2-buten, Sigma-Aldrich®
1,4-Dicyano-2-butene
Formel: C₆H₆N₂
Molecular Weight: 106,1252 g/mol
MDL: MFCD00001964
CAS-Nummer: 1119-85-3
EINECS: 214-287-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR