Bicyclohexyl, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
D79403-25G
D79403-100G
D79403-5G
SIALD79403-25GEA
103
EUR
SIALD79403-25G
SIALD79403-100G
SIALD79403-5G
Bicyclohexyl, Sigma-Aldrich®
Bicyclohexyl
Formel:
C₁₂H₂₂ Molecular Weight: 166.31 g/mol Siedepunkt: 227 Dichte: 0.86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003815 CAS-Nummer: 92-51-3 EINECS: 202-161-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...