(±)-γ-Decalacton, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
(±)-4-Hydroxydecansäurelacton, (±)-γ-Dekalacton, (±)-Decan-4-olid
M-Clarity™ Qualitätsstufe = MQ100
D804-5G
SIALD804-5GEA
33.3
EUR
SIALD804-5G
(±)-γ-Decalacton, Sigma-Aldrich®
(±)-γ-Decalacton
Formel:
C₁₀H₁₈O₂ Molecular Weight: 170.25 g/mol Siedepunkt: 281 Dichte: 0.965 g/cm³ (0 °C) |
MDL:
MFCD00005404 CAS-Nummer: 706-14-9 EINECS: 211-892-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...