Diethoxy(diphenyl)silan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diphenyldiethoxysilan

M-Clarity™ Qualitätsstufe = MQ100

D83532-50ML
SIALD83532-50MLEA 127 EUR
SIALD83532-50ML
Diethoxy(diphenyl)silan, Sigma-Aldrich®
Diethoxy(diphenyl)silan
Formel: C₁₆H₂₀O₂Si
Molecular Weight: 272,42 g/mol
MDL: MFCD00015126
CAS-Nummer: 2553-19-7
EINECS: 219-860-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR