Dicloxacillin sodium salt monohydrate 95,0-101,0%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2,6-Dichlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Sodium (2S,5R,6R)-6-(3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Dicloxacillin Sodium hydrate

M-Clarity™ Qualitätsstufe = MQ200

D9016-1G
SIALD9016-1GEA 94.2 EUR
SIALD9016-1G
Dicloxacillin sodium salt monohydrate 95,0-101,0%, Sigma-Aldrich®
Dicloxacillin sodium salt monohydrate
Formel: C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O
Molecular Weight: 510.33 g/mol
MDL: MFCD00056865
CAS-Nummer: 13412-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR