o-Dianisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3'-Dimethoxybenzidin

M-Clarity™ Qualitätsstufe = MQ200

D9143-5G
SIALD9143-5GEA 53.1 EUR
SIALD9143-5G
o-Dianisidin, Sigma-Aldrich®
o-Dianisidin
Formel: C₁₄H₁₆N₂O₂
Molecular Weight: 244,29 g/mol
MDL: MFCD00008372
CAS-Nummer: 119-90-4
EINECS: 204-355-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR