Diethylchlorphosphit, Sigma-Aldrich®
Lieferant: Merck
MQ100
D91659-100G
D91659-25G
D91659-5G
SIALD91659-100GEA
521
EUR
SIALD91659-100G
SIALD91659-25G
SIALD91659-5G
Diethylchlorphosphit, Sigma-Aldrich®
Diethylchlorphosphit
Formel:
C₄H₁₀ClO₂P Molecular Weight: 156.55 g/mol |
MDL:
MFCD00009074 CAS-Nummer: 589-57-1 EINECS: 209-652-2 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...