Diethylchlorphosphit, Sigma-Aldrich®

Lieferant: Merck


MQ100

D91659-100G D91659-25G D91659-5G
SIALD91659-100GEA 521 EUR
SIALD91659-100G SIALD91659-25G SIALD91659-5G
Diethylchlorphosphit, Sigma-Aldrich®
Diethylchlorphosphit
Formel: C₄H₁₀ClO₂P
Molecular Weight: 156.55 g/mol
MDL: MFCD00009074
CAS-Nummer: 589-57-1
EINECS: 209-652-2
UN: 2920
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR