Diethylethylmalonat, Sigma-Aldrich®

Lieferant: Merck


MQ100

D95204-100G
SIALD95204-100GEA 50.8 EUR
SIALD95204-100G
Diethylethylmalonat, Sigma-Aldrich®
Diethylethylmalonat
Formel: C₉H₁₆O₄
Molecular Weight: 188.22 g/mol
MDL: MFCD00009167
CAS-Nummer: 133-13-1
EINECS: 205-093-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR