3-(3,4-Dihydroxyphenyl)-L-alanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

MQ200

D9628-100G D9628-25G D9628-500G D9628-5G
SIALD9628-100GEA 805 EUR
SIALD9628-100G SIALD9628-25G SIALD9628-500G SIALD9628-5G
3-(3,4-Dihydroxyphenyl)-L-alanin, Sigma-Aldrich®
3-(3,4-Dihydroxyphenyl)-L-alanin
Formel: C₉H₁₁NO₄
Molecular Weight: 197.19 g/mol
MDL: MFCD00002598
CAS-Nummer: 59-92-7
EINECS: 200-445-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR