3-(3,4-Dihydroxyphenyl)-L-alanin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
MQ200
D9628-100G
D9628-25G
D9628-500G
D9628-5G
SIALD9628-100GEA
805
EUR
SIALD9628-100G
SIALD9628-25G
SIALD9628-500G
SIALD9628-5G
3-(3,4-Dihydroxyphenyl)-L-alanin, Sigma-Aldrich®
3-(3,4-Dihydroxyphenyl)-L-alanin
Formel:
C₉H₁₁NO₄ Molecular Weight: 197.19 g/mol |
MDL:
MFCD00002598 CAS-Nummer: 59-92-7 EINECS: 200-445-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...