Diethyl(2,2-diethoxyethyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 2,2-diethoxy-ethanephosphonate

M-Clarity™ Qualitätsstufe = MQ200

D99250-25G
SIALD99250-25GEA 231 EUR
SIALD99250-25G
Diethyl(2,2-diethoxyethyl)phosphonat, Sigma-Aldrich®
Diethyl(2,2-diethoxyethyl)phosphonat
Formel: C₁₀H₂₃O₅P
Molecular Weight: 254,26 g/mol
MDL: MFCD00009236
CAS-Nummer: 7598-61-0
EINECS: 231-504-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR