Density standards, determined at 15 to 150 °C, Supelco®
Lieferant: Merck
DEN10001-60ML DEN10002-60ML DEN15001-60ML DEN15002-60ML DEN1501-60ML DEN1504-60ML DEN1506-60ML DEN1507-60ML DEN1508-60ML DEN1510-60ML DEN1512-60ML DEN2001-60ML DEN2002-60ML DEN2003-60ML DEN2004-60ML DEN2006-60ML DEN2008-60ML DEN2009-60ML DEN2010-60ML DEN2011-60ML DEN2012-60ML DEN2013-60ML DEN2014-60ML DEN2502-60ML DEN2503-60ML DEN2507-60ML DEN2508-60ML DEN2509-60ML DEN2510-60ML DEN4001-60ML DEN4003-60ML DEN4005-60ML DEN5001-60ML DEN5002-60ML DEN5003-60ML DEN6001-60ML DEN6002-60ML DEN6003-60ML DEN8003-60ML
SIALDEN10001-60MLEA 89.2 EUR
SIALDEN10001-60ML SIALDEN10002-60ML SIALDEN15001-60ML SIALDEN15002-60ML SIALDEN1501-60ML SIALDEN1504-60ML SIALDEN1506-60ML SIALDEN1507-60ML SIALDEN1508-60ML SIALDEN1510-60ML SIALDEN1512-60ML SIALDEN2001-60ML SIALDEN2002-60ML SIALDEN2003-60ML SIALDEN2004-60ML SIALDEN2006-60ML SIALDEN2008-60ML SIALDEN2009-60ML SIALDEN2010-60ML SIALDEN2011-60ML SIALDEN2012-60ML SIALDEN2013-60ML SIALDEN2014-60ML SIALDEN2502-60ML SIALDEN2503-60ML SIALDEN2507-60ML SIALDEN2508-60ML SIALDEN2509-60ML SIALDEN2510-60ML SIALDEN4001-60ML SIALDEN4003-60ML SIALDEN4005-60ML SIALDEN5001-60ML SIALDEN5002-60ML SIALDEN5003-60ML SIALDEN6001-60ML SIALDEN6002-60ML SIALDEN6003-60ML SIALDEN8003-60ML
Density standards, determined at 15 to 150 °C, Supelco®
Standards Dichtestandards
  • Dual certified to ISO 17025 / ISO 17034 under UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
Standards: These products are certified reference materials (CRMs) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Bestellinformation: Supplied in tamper evident packaging (please refer to standard for shelf life).