Density standards, determined at 15 to 150 °C, Supelco®
Lieferant: Merck
DEN10001-60ML
DEN10002-60ML
DEN15001-60ML
DEN15002-60ML
DEN1501-60ML
DEN1504-60ML
DEN1506-60ML
DEN1507-60ML
DEN1508-60ML
DEN1510-60ML
DEN1512-60ML
DEN2001-60ML
DEN2002-60ML
DEN2003-60ML
DEN2004-60ML
DEN2006-60ML
DEN2008-60ML
DEN2009-60ML
DEN2010-60ML
DEN2011-60ML
DEN2012-60ML
DEN2013-60ML
DEN2014-60ML
DEN2502-60ML
DEN2503-60ML
DEN2507-60ML
DEN2508-60ML
DEN2509-60ML
DEN2510-60ML
DEN4001-60ML
DEN4003-60ML
DEN4005-60ML
DEN5001-60ML
DEN5002-60ML
DEN5003-60ML
DEN6001-60ML
DEN6002-60ML
DEN6003-60ML
DEN8003-60ML
SIALDEN10001-60MLEA
89.2
EUR
SIALDEN10001-60ML
SIALDEN10002-60ML
SIALDEN15001-60ML
SIALDEN15002-60ML
SIALDEN1501-60ML
SIALDEN1504-60ML
SIALDEN1506-60ML
SIALDEN1507-60ML
SIALDEN1508-60ML
SIALDEN1510-60ML
SIALDEN1512-60ML
SIALDEN2001-60ML
SIALDEN2002-60ML
SIALDEN2003-60ML
SIALDEN2004-60ML
SIALDEN2006-60ML
SIALDEN2008-60ML
SIALDEN2009-60ML
SIALDEN2010-60ML
SIALDEN2011-60ML
SIALDEN2012-60ML
SIALDEN2013-60ML
SIALDEN2014-60ML
SIALDEN2502-60ML
SIALDEN2503-60ML
SIALDEN2507-60ML
SIALDEN2508-60ML
SIALDEN2509-60ML
SIALDEN2510-60ML
SIALDEN4001-60ML
SIALDEN4003-60ML
SIALDEN4005-60ML
SIALDEN5001-60ML
SIALDEN5002-60ML
SIALDEN5003-60ML
SIALDEN6001-60ML
SIALDEN6002-60ML
SIALDEN6003-60ML
SIALDEN8003-60ML
Density standards, determined at 15 to 150 °C, Supelco®
Standards
Dichtestandards
- Dual certified to ISO 17025 / ISO 17034 under UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols
Standards: These products are certified reference materials (CRMs) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Bestellinformation: Supplied in tamper evident packaging (please refer to standard for shelf life).