EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

MQ100

E103-1G
SIALE103-1GEA 13.2 EUR
SIALE103-1G
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat), Sigma-Aldrich®
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247.29 g/mol
MDL: MFCD00006703
CAS-Nummer: 16357-59-8
EINECS: 240-418-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR