(±)-Ethyl-2-brompropionat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate
M-Clarity™ Qualitätsstufe = MQ200
E14551-100G
E14551-250G
E14551-25G
SIALE14551-100GEA
48.2
EUR
SIALE14551-100G
SIALE14551-250G
SIALE14551-25G
(±)-Ethyl-2-brompropionat, Sigma-Aldrich®
(±)-Ethyl-2-brompropionat
Formel:
C₅H₉BrO₂ |
MDL:
MFCD00000144 CAS-Nummer: 535-11-5 EINECS: 208-609-5 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...