Ethylcarbazat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl hydrazinecarboxylate

M-Clarity™ Qualitätsstufe = MQ200

E16503-25G
SIALE16503-25GEA 60.5 EUR
SIALE16503-25G
Ethylcarbazat, Sigma-Aldrich®
Ethylcarbazat
Formel: C₃H₈N₂O₂
Molecular Weight: 104,11 g/mol
MDL: MFCD00007595
CAS-Nummer: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR