Ethyl-4-chlorbutyrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

E16953-100G
SIALE16953-100GEA 75.9 EUR
SIALE16953-100G
Ethyl-4-chlorbutyrat, Sigma-Aldrich®
Ethyl-4-chlorbutyrat
Formel: C₆H₁₁ClO₂
Molecular Weight: 150,6 g/mol
MDL: MFCD00001004
CAS-Nummer: 3153-36-4
EINECS: 221-591-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR