, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid

M-Clarity™ Qualitätsstufe = MQ400

E1894-100G E1894-1KG E1894-250G E1894-25G
SIALE1894-100GEA 489 EUR
SIALE1894-100G SIALE1894-1KG SIALE1894-250G SIALE1894-25G
, Sigma-Aldrich®
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)
Formel: C₉H₂₀N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00006160
CAS-Nummer: 16052-06-5
EINECS: 240-198-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR