Ethylcyclopropancarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

E21604-25G
SIALE21604-25GEA 103 EUR
SIALE21604-25G
Ethylcyclopropancarboxylat, Sigma-Aldrich®
Ethylcyclopropancarboxylat
Formel: C₆H₁₀O₂
Molecular Weight: 114,15 g/mol
MDL: MFCD00001282
CAS-Nummer: 4606-07-9
EINECS: 225-010-4
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR