Ethyl-2-furoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl2-furoate, Ethyl furan-2-carboxylate
M-Clarity™ Qualitätsstufe = MQ100
E28501-100G
SIALE28501-100GEA
77.2
EUR
SIALE28501-100G
Ethyl-2-furoat, Sigma-Aldrich®
Ethyl-2-furoat
Formel:
C₇H₈O₃ Molecular Weight: 140,14 g/mol |
MDL:
MFCD00003237 CAS-Nummer: 614-99-3 EINECS: 210-404-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...