Ethyl-2-indolcarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

M-Clarity™ Qualitätsstufe = MQ100

E32800-25G E32800-5G
SIALE32800-25GEA 152 EUR
SIALE32800-25G SIALE32800-5G
Ethyl-2-indolcarboxylat, Sigma-Aldrich®
Ethyl-2-indolcarboxylat
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189.21 g/mol
MDL: MFCD00005609
CAS-Nummer: 3770-50-1
EINECS: 223-206-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR