Ethylisobutyrylacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 4-methyl-3-oxopentanoate

M-Clarity™ Qualitätsstufe = MQ200

E33203-10G
SIALE33203-10GEA 52 EUR
SIALE33203-10G
Ethylisobutyrylacetat, Sigma-Aldrich®
Ethylisobutyrylacetat
Formel: C₈H₁₄O₃
Molecular Weight: 158,2 g/mol
MDL: MFCD00009198
CAS-Nummer: 7152-15-0
EINECS: 230-491-9
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR