Ethylpicolinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate

M-Clarity™ Qualitätsstufe = MQ200

E45414-25G
SIALE45414-25GEA 79.3 EUR
SIALE45414-25G
Ethylpicolinat, Sigma-Aldrich®
Ethylpicolinat
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00006292
CAS-Nummer: 2524-52-9
EINECS: 219-758-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR