EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
E8145-10G
E8145-50G
SIALE8145-10GEA
146
EUR
SIALE8145-10G
SIALE8145-50G
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz), Sigma-Aldrich®
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz)
Formel:
C₁₄H₂₀N₂Na₄O₁₀ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274420 CAS-Nummer: 13368-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...