EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure), Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ400

E9502-100G E9502-10G E9502-1KG E9502-250G E9502-25G E9502-5KG
SIALE9502-100GEA 151 EUR
SIALE9502-100G SIALE9502-10G SIALE9502-1KG SIALE9502-250G SIALE9502-25G SIALE9502-5KG
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure), Sigma-Aldrich®
Formel: C₉H₂₀N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00006160
CAS-Nummer: 16052-06-5
EINECS: 240-198-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR