5-Fluor-2-indolcarbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Fluoro-1H-indole-2-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
F109-250MG
SIALF109-250MGEA
61.2
EUR
SIALF109-250MG
5-Fluor-2-indolcarbonsäure, Sigma-Aldrich®
5-Fluoroindole-2-carboxylic acid
Formel:
C₉H₆FNO₂ Schmelzpunkt: 258…259 °C |
MDL:
MFCD00005612 CAS-Nummer: 399-76-8 EINECS: 206-919-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...