1-Fluorencarbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
F1204-1G
SIALF1204-1GEA
44.2
EUR
SIALF1204-1G
1-Fluorencarbonsäure, Sigma-Aldrich®
1-Fluorencarbonsäure
Formel:
C₁₄H₁₀O₂ Molecular Weight: 210,23 g/mol |
MDL:
MFCD00001130 CAS-Nummer: 6276-03-5 EINECS: 228-472-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...