Bis(4-fluorphenyl)sulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-Difluordiphenylsulfon, 4-Fluorphenylsulfon

M-Clarity™ Qualitätsstufe = MQ100

F15145-25G
SIALF15145-25GEA 141 EUR
SIALF15145-25G
Bis(4-fluorphenyl)sulfon, Sigma-Aldrich®
Bis(4-fluorphenyl)sulfon
Formel: C₁₂H₈F₂O₂S
Molecular Weight: 254,26 g/mol
MDL: MFCD00000350
CAS-Nummer: 383-29-9
EINECS: 206-847-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR