3-(2-Furyl)acrylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Furan-2-yl)acrylic acid

M-Clarity™ Qualitätsstufe = MQ100

F20807-25G
SIALF20807-25GEA 61.8 EUR
SIALF20807-25G
3-(2-Furyl)acrylsäure, Sigma-Aldrich®
3-(2-Furyl)acrylsäure
Formel: C₇H₆O₃
Molecular Weight: 138,12 g/mol
MDL: MFCD00003257
CAS-Nummer: 539-47-9
EINECS: 208-718-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR