Ficoll® 400, BioXtra, lyophilised powder, for molecular biology, Sigma-Aldrich®
Lieferant: Merck
F2637-10G F2637-100G F2637-25G F2637-5G
SIALF2637-10GEA 129 EUR
SIALF2637-10G SIALF2637-100G SIALF2637-25G SIALF2637-5G
Ficoll® 400, BioXtra, lyophilised powder, for molecular biology, Sigma-Aldrich®
Zelltrennmedien
A non-ionic synthetic polymer of sucrose.
  • Non-ionic polymer free of DNases, RNases, phosphatases and proteases
  • Used for cell separation and organ isolation
  • Separates cells with similar density but different size (PBMC from human peripheral blood, bone marrow and umbilical cord blood)
  • Density ranges attained up to 1,2 g/ml
Ficoll® 400 polymer is known as synthetic polymer of sucrose which produce density gradients to separate subcellular components.