5-Fluortryptamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(5-Fluoro-1H-indol-3-yl)ethanamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

F7001-250MG
SIALF7001-250MGEA 301 EUR
SIALF7001-250MG
5-Fluortryptamin Hydrochlorid, Sigma-Aldrich®
5-Fluortryptamin Hydrochlorid
Formel: C₁₀H₁₂ClFN₂
Molecular Weight: 214.67 g/mol
MDL: MFCD00051081
CAS-Nummer: 2711-58-2
EINECS: 220-307-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR